(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥98%, ee 99%

Supplier: Ambeed
Danger

Synonyms: (2R)-(-)-Glycidyl p-toluenesulphonate, (2R)-(-)-Glycidyl tosylate

A167572-10G A167572-1G A167572-5G
76690-084EA 21.22 USD
76690-084 76690-086 76690-088
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥98%, ee 99%
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄S
MW: 228.2649 g/mol
MDL Number: MFCD00010834
CAS Number: 113826-06-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR