2-Bromo-5-hydroxy-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A166435-100G A166435-10G A166435-1G A166435-25G A166435-500G A166435-5G
76690-060EA 129.43 USD
76690-060 76690-062 76690-064 76690-066 76690-068 76690-070
2-Bromo-5-hydroxy-4-methoxybenzaldehyde 98%
2-Bromo-5-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00195552
CAS Number: 2973-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR