Butyl octyl phthalate 95%

Supplier: Ambeed
Warning

A166017-1G A166017-250MG
76690-032EA 181.42 USD
76690-032 76690-034
Butyl octyl phthalate 95%
Butyl octyl phthalate

Precaución: for Research Use Only

Formula: C₂₀H₃₀O₄
MW: 334.45 g/mol
MDL Number: MFCD00072261
CAS Number: 84-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR