1,3-Diphenylurea 95%

Supplier: Ambeed
Warning

A165076-1000G A165076-100G A165076-25G A165076-500G
76689-992EA 35 USD
76689-992 76689-994 76689-996 76689-998
1,3-Diphenylurea 95%
1,3-Diphenylurea

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MDL Number: MFCD00003017
CAS Number: 102-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR