4'-Methyl-2,2,2-trifluoroacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-1-(p-tolyl)ethanone, 4-(Trifluoroacetyl)toluene, 2,2,2-Trifluoro-1-(4-methylphenyl)ethan-1-one

A164101-10G A164101-1G A164101-25G A164101-5G
76689-944EA 65.78 USD
76689-944 76689-946 76689-948 76689-950
4'-Methyl-2,2,2-trifluoroacetophenone 95%
4'-Methyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MW: 188.15 g/mol
MDL Number: MFCD00052338
CAS Number: 394-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR