Butyl valerate 95%

Supplier: Ambeed
Warning

A163558-100G A163558-25G A163558-500G A163558-5G
76689-912EA 60 USD
76689-912 76689-914 76689-916 76689-918
Butyl valerate 95%
Butyl valerate

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MDL Number: MFCD00015258
CAS Number: 591-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR