7-Amino-1,8-naphthyridin-2(8H)-one 97%

Supplier: Ambeed
Warning

A162993-100MG A162993-10G A162993-1G A162993-250MG A162993-5G
76689-888EA 7.43 USD
76689-888 76689-890 76689-892 76689-894 76689-896
7-Amino-1,8-naphthyridin-2(8H)-one 97%
7-Amino-1,8-naphthyridin-2(8H)-one

Precaución: for Research Use Only

Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD09743308
CAS Number: 1931-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR