7-Bromo-5-nitroindole 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-5-nitro-1H-indole

A162321-1G A162321-250MG A162321-5G
76689-870EA 177.18 USD
76689-870 76689-872 76689-874
7-Bromo-5-nitroindole 95%
7-Bromo-5-nitroindole

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.05 g/mol
MDL Number: MFCD04038858
CAS Number: 87240-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR