7-Amino-4-methylquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A162218-100MG A162218-1G A162218-250MG A162218-5G
76689-862EA 14.86 USD
76689-862 76689-864 76689-866 76689-868
7-Amino-4-methylquinolin-2(1H)-one 97%
7-Amino-4-methylquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.19 g/mol
MDL Number: MFCD00134256
CAS Number: 19840-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR