Allyldiphenylphosphine oxide 98%

Supplier: Ambeed
Danger

A162193-10G A162193-250MG A162193-5G A162193-1G
76689-856EA 409 USD
76689-856 76689-858 76689-860 77600-300
Allyldiphenylphosphine oxide 98%
Allyldiphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₅H₁₅OP
MDL Number: MFCD00013908
CAS Number: 4141-48-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR