N-Boc-cyclopropylamine 97%

Supplier: Ambeed
Warning

A161433-100G A161433-10G A161433-1G A161433-5G A161433-25G
76689-844EA 231 USD
76689-844 76689-846 76689-848 77625-560 77625-558
N-Boc-cyclopropylamine 97%
N-Boc-cyclopropylamine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD09701238
CAS Number: 132844-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR