Benzyl N-(2-aminoethyl)carbamate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A161038-100G
A161038-10G
A161038-1G
A161038-250MG
A161038-5G
A161038-25G
76689-820EA
239
USD
76689-820
76689-822
76689-824
76689-826
76689-828
77567-348
Benzyl N-(2-aminoethyl)carbamate hydrochloride 97%
Benzyl N-(2-aminoethyl)carbamate hydrochloride
Precaución: For research use only.
Formula:
C₁₀H₁₄N₂O₂ |
MDL Number:
MFCD00270150 CAS Number: 18807-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...