Bethanechol chloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Carbamoyloxy)-N,N,N-trimethyl-1-propanaminium Chloride

A160422-1G A160422-250MG
76689-816EA 9.55 USD
76689-816 76689-818
Bethanechol chloride 98%
Bethanechol chloride

Precaución: for Research Use Only

Formula: C₇H₁₇ClN₂O₂
MW: 196.675 g/mol
MDL Number: MFCD00055224
CAS Number: 590-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR