Methyl-2-methoxy-4-methylbenzoate 97%

Supplier: Ambeed
Warning

A159937-1G A159937-250MG A159937-25G A159937-5G
76689-774EA 11 USD
76689-774 76689-776 76689-778 76689-780
Methyl-2-methoxy-4-methylbenzoate 97%
Methyl-2-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD07782177
CAS Number: 81245-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR