Chloromethyl ethyl carbonate 97%

Supplier: Ambeed
Danger

A159896-100MG A159896-1G A159896-250MG A159896-25G A159896-5G A159896-10G
76689-764EA 5 USD
76689-764 76689-766 76689-768 76689-770 76689-772 77600-266
Chloromethyl ethyl carbonate 97%
Chloromethyl ethyl carbonate

Precaución: for Research Use Only

Formula: C₄H₇ClO₃
MDL Number: MFCD00137995
CAS Number: 35179-98-7
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR