4,5-Dimethoxy-2-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

A159638-100G A159638-10G A159638-1G A159638-25G A159638-500G A159638-5G
76689-752EA 147 USD
76689-752 76689-754 76689-756 76689-758 76689-760 76689-762
4,5-Dimethoxy-2-nitrobenzyl alcohol 98%
4,5-Dimethoxy-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁NO₅
MDL Number: MFCD00014701
CAS Number: 1016-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR