Boc-D-Arg-OH·HCl·H2O 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Arg-OH·HCl monohydrate, (R)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid hydrochloride hydrate, Nα-Boc-D-arginine hydrochloride monohydrate, Nα-(tert-Butoxycarbonyl)-D-arginine hydrochloride monohydrate

A159002-100G A159002-10G A159002-1G A159002-250MG A159002-25G A159002-5G
76689-724EA 195.21 USD
76689-724 76689-726 76689-728 76689-730 76689-732 76689-734
Boc-D-Arg-OH·HCl·H2O 95%
Boc-D-Arg-OH·HCl·H2O

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₄O₄·HCl·H₂O
MW: 328.80 g/mol
MDL Number: MFCD00065554
CAS Number: 204070-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR