(3-Bromo-4-methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 2-(3-Bromo-4-methoxyphenyl)acetonitrile

A158948-10G A158948-25G A158948-5G
76689-718EA 76.39 USD
76689-718 76689-720 76689-722
(3-Bromo-4-methoxyphenyl)acetonitrile 98%
(3-Bromo-4-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.08 g/mol
MDL Number: MFCD00016391
CAS Number: 772-59-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR