Acrivastine 95%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(6-((E)-3-(Pyrrolidin-1-yl)-1-(p-tolyl)prop-1-en-1-yl)pyridin-2-yl)acrylic acid

A158908-10MG A158908-1MG A158908-50MG
76689-712EA 80.63 USD
76689-712 76689-714 76689-716
Acrivastine 95%
Acrivastine

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₂O₂
MW: 348.45 g/mol
MDL Number: MFCD00869830
CAS Number: 87848-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR