8-Bromo-5,6-difluoro-2-methylquinoline 97%

Supplier: Ambeed
Warning

A158863-1G A158863-5G
76689-708EA 31.83 USD
76689-708 76689-710
8-Bromo-5,6-difluoro-2-methylquinoline 97%
8-Bromo-5,6-difluoro-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₂N
MW: 258.06 g/mol
MDL Number: MFCD08705815
CAS Number: 131190-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR