Dantrolene sodium hemiheptahydrate 98%

Supplier: Ambeed
Warning

Synonyms: 1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione, sodium salt hydrate(2:2:7), Sodium Dantrolene, Dantrolene, 1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt, Dantrium

A158810-100MG A158810-1G A158810-1MG A158810-5G
76689-706EA 6 USD
76689-706 77600-260 77600-262 77879-091
Dantrolene sodium hemiheptahydrate 98%
Dantrolene sodium hemiheptahydrate

Precaución: for Research Use Only

Formula: C₂₈H₃₄N₈Na₂O₁₇
MDL Number: MFCD07357270
CAS Number: 24868-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR