4-(1,3-Benzothiazol-2-yl)phenylamine 97%

Supplier: Ambeed
Warning

A158766-100G A158766-25G A158766-5G
76689-700EA 185 USD
76689-700 76689-702 76689-704
4-(1,3-Benzothiazol-2-yl)phenylamine 97%
4-(1,3-Benzothiazol-2-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂S
MDL Number: MFCD00182896
CAS Number: 6278-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR