4-Nitro-D-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Phe(4-NO2)-OH, H-D-Phe(4-NO₂)-OH
A158172-10G
A158172-5G
A158172-25G
A158172-100G
76689-680EA
17
USD
76689-680
76689-682
77600-250
77625-468
4-Nitro-D-phenylalanine 98%
4-Nitro-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂O₄ |
MDL Number:
MFCD00066016 CAS Number: 56613-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...