(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (2S,4R)-4-hydroxy-2-hydroxymethylpyrrolidine-1-carboxylate, N-Boc-trans-4-hydroxy-L-prolinol, (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine, Boc-Hyp-OL
A158147-10G
A158147-1G
A158147-250MG
A158147-25G
A158147-5G
A158147-100G
76689-670EA
37
USD
76689-670
76689-672
76689-674
76689-676
76689-678
77600-248
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 97%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₄ |
MDL Number:
MFCD02094386 CAS Number: 61478-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...