4-Bromo-5-methylisatin 97%

Supplier: Ambeed
Warning

A157940-100G A157940-10G A157940-1G A157940-25G A157940-5G
76689-654EA 135 USD
76689-654 76689-656 76689-658 76689-660 76689-662
4-Bromo-5-methylisatin 97%
4-Bromo-5-methylisatin

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD00560709
CAS Number: 147149-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR