7-Fluoroquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-4-hydroxyquinazoline, 7-Fluoro-4-quinazolinone, 7-Fluoro-4-quinazolinol

A157905-100G A157905-10G A157905-1G A157905-25G A157905-5G
76689-642EA 195 USD
76689-642 76689-644 76689-646 76689-648 76689-650 76689-652
7-Fluoroquinazolin-4(3H)-one 95%
7-Fluoroquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MDL Number: MFCD05664874
CAS Number: 16499-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR