Bis(2-ethylhexyl)sebacate 97%

Supplier: Ambeed
Warning

Synonyms: 1,10-Decanedioic acid di(2-ethylhexyl) ester, Sebacic acid di(2-ethylhexyl) ester

A157689-10G A157689-25G A157689-1000G
76689-638EA 6 USD
76689-638 76689-640 77877-043
Bis(2-ethylhexyl)sebacate 97%
Bis(2-ethylhexyl)sebacate

Precaución: for Research Use Only

Formula: C₂₆H₅₀O₄
MW: 426.68 g/mol
Boiling Pt: 435
Melting Pt: -55
Flash Pt: 213
MDL Number: MFCD00009497
CAS Number: 122-62-3
Merck Index: 13,01251

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR