7-Chlorothieno[2,3-c]pyridine 95%

Supplier: Ambeed
Warning

A157600-100MG A157600-1G A157600-250MG A157600-5G
76689-630EA 32.89 USD
76689-630 76689-632 76689-634 76689-636
7-Chlorothieno[2,3-c]pyridine 95%
7-Chlorothieno[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MW: 169.631 g/mol
MDL Number: MFCD04973300
CAS Number: 28948-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR