5-Acetyl-2,3-dihydrobenzo[b]furan 95%

Supplier: Ambeed
Warning

A157561-1G A157561-250MG
76689-626EA 16.98 USD
76689-626 76689-628
5-Acetyl-2,3-dihydrobenzo[b]furan 95%
5-Acetyl-2,3-dihydrobenzo[b]furan

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00068033
CAS Number: 90843-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR