7-(4-(4-(2,3-Dichlorophenyl)piperazin-1-yl)butoxy)quinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

Synonyms: Dehydroaripiprazole

A156606-100MG A156606-250MG A156606-1G
76689-588EA 128.37 USD
76689-588 76689-590 77625-442
7-(4-(4-(2,3-Dichlorophenyl)piperazin-1-yl)butoxy)quinolin-2(1H)-one 97%
7-(4-(4-(2,3-Dichlorophenyl)piperazin-1-yl)butoxy)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₂₃H₂₅Cl₂N₃O₂
MW: 446.37 g/mol
MDL Number: MFCD00892073
CAS Number: 129722-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR