2,3-Dioxoindoline-7-carboxylic acid 96%

Supplier: Ambeed
Warning

A156543-1G A156543-250MG A156543-25G A156543-5G
76689-580EA 26 USD
76689-580 76689-582 76689-584 76689-586
2,3-Dioxoindoline-7-carboxylic acid 96%
2,3-Dioxoindoline-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅NO₄
MDL Number: MFCD00612492
CAS Number: 25128-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR