5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate 95%

Supplier: Ambeed
Warning

A154917-100G A154917-1G A154917-25G A154917-500G A154917-5G
76689-524EA 493 USD
76689-524 76689-526 76689-528 76689-530 76689-532
5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate 95%
5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₆S
MDL Number: MFCD00467095
CAS Number: 83763-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR