8-Methoxychroman-4-one 98%

Supplier: Ambeed
Warning

A154905-100MG A154905-1G A154905-250MG A154905-5G
76689-516EA 14 USD
76689-516 76689-518 76689-520 76689-522
8-Methoxychroman-4-one 98%
8-Methoxychroman-4-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD00094977
CAS Number: 20351-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR