(R)-Glycidyl-3-nitrobenzenesulfonate 97%

Supplier: Ambeed
Danger

Synonyms: (R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate

A154692-10G A154692-1G A154692-5G
76689-502EA 35.01 USD
76689-502 76689-504 76689-506
(R)-Glycidyl-3-nitrobenzenesulfonate 97%
(R)-Glycidyl-3-nitrobenzenesulfonate

Precaución: for Research Use Only

Formula: C₉H₉NO₆S
MW: 259.23 g/mol
MDL Number: MFCD00013436
CAS Number: 115314-17-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR