1-[(4-Methylphenyl)sulfonyl]acetone 95%

Supplier: Ambeed
Warning

Synonyms: 1-Tosylpropan-2-one

A154513-25G A154513-5G
76689-496EA 103.97 USD
76689-496 76689-498
1-[(4-Methylphenyl)sulfonyl]acetone 95%
1-[(4-Methylphenyl)sulfonyl]acetone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃S
MW: 212.27 g/mol
MDL Number: MFCD00026240
CAS Number: 5366-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR