6-(tert-Butyl)naphthalene-2,3-dicarbonitrile 95%

Supplier: Ambeed
Warning

A154438-100MG A154438-1G A154438-250MG A154438-5G
76689-484EA 30.77 USD
76689-484 76689-486 76689-488 76689-490
6-(tert-Butyl)naphthalene-2,3-dicarbonitrile 95%
6-(tert-Butyl)naphthalene-2,3-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂
MW: 234.29 g/mol
MDL Number: MFCD00209558
CAS Number: 32703-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR