4-(tert-Butyl)-2-nitroaniline 97%

Supplier: Ambeed
Warning

A153800-100G A153800-10G A153800-1G A153800-25G A153800-5G
76689-460EA 181 USD
76689-460 76689-462 76689-464 76689-466 76689-468
4-(tert-Butyl)-2-nitroaniline 97%
4-(tert-Butyl)-2-nitroaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD00066305
CAS Number: 6310-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR