Ac-β-Ala-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
A152946-100G
A152946-10G
A152946-1G
A152946-25G
A152946-5G
76689-440EA
129.43
USD
76689-440
76689-442
76689-444
76689-446
76689-448
Ac-β-Ala-OH 97%
Ac-β-Ala-OH
Precaución: for Research Use Only
Formula:
C₅H₉NO₃ MW: 131.1 g/mol |
MDL Number:
MFCD00037296 CAS Number: 3025-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...