Boc-D-Asn(Trt)-OH 97%

Supplier: Ambeed
Warning

A152696-100MG A152696-5G
76689-412EA 5 USD
76689-412 76689-414 76689-416 76689-418
Boc-D-Asn(Trt)-OH 97%
Boc-D-Asn(Trt)-OH

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₂O₅
MDL Number: MFCD00153300
CAS Number: 210529-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR