4-Fluorophenethyl bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene

A152452-10G A152452-1G A152452-25G A152452-5G A152452-100G
76689-400EA 22 USD
76689-400 76689-402 76689-404 77549-918 77877-015
4-Fluorophenethyl bromide 98%
4-Fluorophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD03844794
CAS Number: 332-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR