α-D-Galacturonic acid monohydrate 95%

Supplier: Ambeed
Warning

A152008-10G A152008-1G A152008-25G A152008-5G
76689-392EA 93 USD
76689-392 76689-394 76689-396 76689-398
α-D-Galacturonic acid monohydrate 95%
alpha-D-Galacturonic acid monohydrate

Precaución: for Research Use Only

Formula: C₆H₁₀O₇·H₂O
MDL Number: MFCD00071585
CAS Number: 91510-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR