Cytidine-5'-diphosphocholine sodium salt (CDP choline) 97%

Supplier: Ambeed
Warning

Synonyms: CDP-Choline, 2-(((((((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)-N,N,N-trimethylethan-1-aminium, sodium salt(1:1), Citicoline sodium salt, CDP-choline sodium salt

A151969-100G A151969-100MG A151969-5G A151969-25G A151969-10G A151969-1G A151969-500G
76689-384EA 309 USD
76689-384 76689-386 76689-388 77600-154 77624-890 77624-892 77879-056
Cytidine-5'-diphosphocholine sodium salt (CDP choline) 97%
Cytidine-5'-diphosphocholine sodium salt (CDP choline)

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₄NaO₁₁P₂
MW: 510.31 g/mol
MDL Number: MFCD00037486
CAS Number: 33818-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR