6-Bromo-1,4-benzodioxane 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2,3-dihydro-1,4-benzodioxine, 6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

A151598-100G A151598-10G A151598-1G A151598-25G A151598-5G
76689-370EA 216.43 USD
76689-370 76689-372 76689-374 76689-376 76689-378
6-Bromo-1,4-benzodioxane 97%
6-Bromo-1,4-benzodioxane

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD00040750
CAS Number: 52287-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR