Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate
A150761-10G
A150761-1G
A150761-250MG
A150761-25G
A150761-5G
76689-354EA
46.68
USD
76689-354
76689-356
76689-358
76689-360
76689-362
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 97%
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂S MW: 211.28 g/mol |
MDL Number:
MFCD00130099 CAS Number: 108354-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...