7-Bromo-2-methylquinazoline 97%

Supplier: Ambeed
Warning

A150723-100MG A150723-250MG A150723-1G A150723-5G
76689-350EA 34 USD
76689-350 76689-352 77592-832 77624-750
7-Bromo-2-methylquinazoline 97%
7-Bromo-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD11044811
CAS Number: 552331-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR