1,1-Cyclohexanediacetic acid monoamide 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-(2-Amino-2-oxoethyl)cyclohexyl)acetic acid, 2-[1-(2-Amino-2-oxoethyl)cyclohexyl]acetic Acid

A150555-100G A150555-25G A150555-5G
76689-344EA 50.93 USD
76689-344 76689-346 76689-348
1,1-Cyclohexanediacetic acid monoamide 98%
1,1-Cyclohexanediacetic acid monoamide

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.2500 g/mol
MDL Number: MFCD02181086
CAS Number: 99189-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR