(S)-2-(4-Hydroxyphenyl)-7-methoxy-6-(3-methylbut-2-en-1-yl)chroman-4-one 98%

Supplier: Ambeed
Warning

Synonyms: Bavachinin

A150129-1MG A150129-5MG A150129-100MG
76689-336EA 7 USD
76689-336 76689-338 77877-011
(S)-2-(4-Hydroxyphenyl)-7-methoxy-6-(3-methylbut-2-en-1-yl)chroman-4-one 98%
(S)-2-(4-Hydroxyphenyl)-7-methoxy-6-(3-methylbut-2-en-1-yl)chroman-4-one

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₄
MDL Number: MFCD06858307
CAS Number: 19879-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR