Ethyl (1-((2-methyl-1,1-dioxido-3-(pyridin-2-ylcarbamoyl)-2H-benzo[e][1,2]thiazin-4-yl)oxy)ethyl) carbonate 98%

Supplier: Ambeed
Warning

Synonyms: Ampiroxicam

A149917-10MG A149917-1MG A149917-50MG A149917-5MG A149917-1G A149917-5G A149917-250MG
76689-328EA 9.55 USD
76689-328 76689-330 76689-332 76689-334 77600-118 77600-120 77624-620
Ethyl (1-((2-methyl-1,1-dioxido-3-(pyridin-2-ylcarbamoyl)-2H-benzo[e][1,2]thiazin-4-yl)oxy)ethyl) carbonate 98%
Ethyl (1-((2-methyl-1,1-dioxido-3-(pyridin-2-ylcarbamoyl)-2H-benzo[e][1,2]thiazin-4-yl)oxy)ethyl) carbonate

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₃O₇S
MW: 447.46 g/mol
MDL Number: MFCD00866079
CAS Number: 99464-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR