(1H,3H)-Quinazoline-2,4-dione 95%

Supplier: Ambeed
Warning

Synonyms: Quinazoline-2,4(1H,3H)-dione, Benzoyleneurea

A149578-100G A149578-10G A149578-25G A149578-500G A149578-5G
76689-318EA 46.68 USD
76689-318 76689-320 76689-322 76689-324 76689-326
(1H,3H)-Quinazoline-2,4-dione 95%
(1H,3H)-Quinazoline-2,4-dione

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00006699
CAS Number: 86-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR