4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine 98%

Supplier: Ambeed
Warning

Synonyms: Apilimod

A149227-10MG A149227-1MG A149227-5MG A149227-50MG
76689-300EA 44.56 USD
76689-300 76689-302 76689-304 77624-516
4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine 98%
4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₆O₂
MW: 418.49 g/mol
MDL Number: MFCD09954108
CAS Number: 541550-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR